Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylphenylhydrazine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A602576-100G A602576-1G A602576-25G A602576-5G
77254-254EA 285.01 USD InStock
77254-254 77254-256 77254-258 77254-260
4-Isopropylphenylhydrazine hydrochloride 98%
4-Isopropylphenylhydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₅ClN₂
MDL Number: MFCD00013461
CAS Number: 118427-29-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty