Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5aR,10bS)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate 97%

Supplier: AMBEED, INC
Danger

A603578-100MG A603578-250MG
77254-262EA 26 USD InStock
77254-262 77254-264
(5aR,10bS)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate 97%
(5aR,10bS)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₈H₁₃BCl₃F₄N₃O
MDL Number: MFCD28053579
CAS Number: 1252006-72-6
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty