Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chlorophenylthio)acetone 98%

Supplier: AMBEED, INC
Danger

A607664-1G A607664-25G A607664-5G
77280-282EA 7 USD InStock
77280-282 77280-284 77280-286
(4-Chlorophenylthio)acetone 98%
(4-Chlorophenylthio)acetone
Precaución: for Research Use Only


Formula: C₉H₉ClOS
MDL Number: MFCD00013644
CAS Number: 25784-83-2
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty