Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide 98%

Supplier: AMBEED, INC
Warning

A607827-1G A607827-250MG A607827-5G
77266-782EA 47 USD InStock
77266-782 77266-784 77266-786
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide 98%
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide
Precaución: for Research Use Only


Formula: C₂₄H₁₂N₄O₄
MW: 420.38 g/mol
MDL Number: MFCD00326861
CAS Number: 34151-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty