Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK-J4 hydrochloride 98%

Supplier: AMBEED, INC
Warning

A608057-10MG A608057-1MG A608057-25MG
77266-790EA 45 USD InStock
77266-790 77266-792 77266-794
GSK-J4 hydrochloride 98%
GSK-J4 hydrochloride
Precaución: for Research Use Only


Formula: C₂₄H₂₇N₅O₂∙HCl
MDL Number: MFCD26142638
CAS Number: 1797983-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty