Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromodifluoroacetate 98%

Supplier: AMBEED, INC
Danger

A608634-5G A608634-25G A608634-100G
77292-824EA 17 USD InStock
77292-824 77551-826 77617-544
Ethyl bromodifluoroacetate 98%
Ethyl bromodifluoroacetate
Precaución: for Research Use Only


Formula: BrF₂CCOOCH₂CH₃
MW: 202.98 g/mol
Boiling Pt: 112
MDL Number: MFCD00042069
CAS Number: 667-27-6
UN: 2924
ADR: 3,8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty