Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzenesulfonic acid 95%

Supplier: AMBEED, INC
Danger

A616229-1G A616229-5G
77292-886EA 47 USD InStock
77292-886 77292-888
4-Bromobenzenesulfonic acid 95%
4-Bromobenzenesulfonic acid
Precaución: for Research Use Only


Formula: C₆H₅BrO₃S
MDL Number: MFCD04972106
CAS Number: 138-36-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty