Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Pyrazol-5-yl)phenol 98%

Supplier: AMBEED, INC
Warning

A616476-100MG A616476-1G A616476-250MG A616476-5G
77242-742EA 6 USD InStock
77242-742 77242-744 77242-746 77242-748
2-(1H-Pyrazol-5-yl)phenol 98%
2-(1H-Pyrazol-5-yl)phenol
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MW: 160.17 g/mol
MDL Number: MFCD05863242
CAS Number: 34810-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty