Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4''-Tri-9-carbazolyltriphenylamine 98%

Supplier: AMBEED, INC
Warning

A617243-1G A617243-5G A617243-250MG
77292-892EA 42 USD InStock
77292-892 77292-894 77749-264
4,4',4''-Tri-9-carbazolyltriphenylamine 98%
4,4',4''-Tri-9-carbazolyltriphenylamine
Precaución: for Research Use Only


Formula: C₅₄H₃₆N₄
MDL Number: MFCD03093250
CAS Number: 139092-78-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty