Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-boc-3,5-diaminobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A618593-1G A618593-250MG
77280-336EA 125 USD InStock
77280-336 77280-338
Di-boc-3,5-diaminobenzoic acid 97%
Di-boc-3,5-diaminobenzoic acid
Precaución: for Research Use Only


Formula: C₁₇H₂₄N₂O₆
MW: 352.39 g/mol
MDL Number: MFCD00270356
CAS Number: 133887-83-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty