Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-carboxyethyl)heptanedioic acid 97%

Supplier: AMBEED, INC
Warning

A620464-100MG A620464-1G A620464-250MG
77242-786EA 43 USD InStock
77242-786 77242-788 77242-790
4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-carboxyethyl)heptanedioic acid 97%
4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-carboxyethyl)heptanedioic acid
Precaución: for Research Use Only


Formula: C₂₅H₂₇NO₈
MDL Number: MFCD16628194
CAS Number: 798576-99-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty