Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-N,N-diphenylaniline 97%

Supplier: AMBEED, INC
Warning

A620990-100MG A620990-1G A620990-250MG
77292-918EA 51 USD InStock
77292-918 77292-920 77292-922
3-Methyl-N,N-diphenylaniline 97%
3-Methyl-N,N-diphenylaniline
Precaución: for Research Use Only


Formula: C₁₉H₁₇N
MDL Number: MFCD00420287
CAS Number: 4316-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty