Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Ethyl 2-dodecanamido-5-guanidinopentanoate hydrochloride 98%

Supplier: AMBEED, INC
Danger

A621851-25G A621851-5G
77254-404EA 21 USD InStock
77254-404 77254-406
(S)-Ethyl 2-dodecanamido-5-guanidinopentanoate hydrochloride 98%
(S)-Ethyl 2-dodecanamido-5-guanidinopentanoate hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₄₁ClN₄O₃
MW: 421.01 g/mol
MDL Number: MFCD11983056
CAS Number: 60372-77-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty