Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethoxypropan-2-amine 97%

Supplier: AMBEED, INC
Danger

A622493-100MG A622493-5G
77254-416EA 23 USD InStock
77254-416 77747-284
1,3-Dimethoxypropan-2-amine 97%
1,3-Dimethoxypropan-2-amine
Precaución: For research use only.


Formula: C₅H₁₃NO₂
MDL Number: MFCD04038441
CAS Number: 78531-29-0
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty