Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyloxy-2-Iodoethane 98%

Supplier: AMBEED, INC
Danger

A624287-1G A624287-250MG A624287-5G A624287-25G
77266-910EA 34 USD InStock
77266-910 77266-912 77266-914 77749-273
1-Benzyloxy-2-Iodoethane 98%
1-Benzyloxy-2-Iodoethane
Precaución: for Research Use Only


Formula: C₉H₁₁IO
MW: 262.09 g/mol
MDL Number: MFCD10574996
CAS Number: 54555-84-9
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty