Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Azabicyclo[2.2.1]heptane 98%

Supplier: AMBEED, INC
Warning

A624370-100MG A624370-250MG A624370-5G A624370-50MG
77266-916EA 90 USD InStock
77266-916 77266-918 77266-920 77747-287
2-Azabicyclo[2.2.1]heptane 98%
2-Azabicyclo[2.2.1]heptane
Precaución: for Research Use Only


Formula: C₆H₁₁N
MW: 97.16 g/mol
MDL Number: MFCD05864542
CAS Number: 279-24-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty