Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylumbelliferyl-alpha-D-galactopyranoside 98%

Supplier: AMBEED, INC
Warning

A627028-100MG A627028-250MG
77292-934EA 96 USD InStock
77292-934 77292-936
4-Methylumbelliferyl-alpha-D-galactopyranoside 98%
4-Methylumbelliferyl-alpha-D-galactopyranoside
Precaución: for Research Use Only


Formula: C₁₆H₁₈O₈
MDL Number: MFCD00063278
CAS Number: 38597-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty