Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroaniline-3-sulfonic acid 98%

Supplier: AMBEED, INC
Warning

A627847-100G A627847-25G A627847-5G
77292-938EA 83 USD InStock
77292-938 77292-940 77292-942
4-Chloroaniline-3-sulfonic acid 98%
4-Chloroaniline-3-sulfonic acid
Precaución: for Research Use Only


Formula: C₆H₆ClNO₃S
MDL Number: MFCD00025296
CAS Number: 88-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty