Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-methylbenzyl Alcohol 97%

Supplier: AMBEED, INC
Warning

A631020-100MG A631020-1G A631020-250MG A631020-5G
77254-488EA 14 USD InStock
77254-488 77254-490 77254-492 77254-494
2-Amino-5-methylbenzyl Alcohol 97%
2-Amino-5-methylbenzyl Alcohol
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MW: 137.17 g/mol
MDL Number: MFCD00074995
CAS Number: 34897-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty