Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1H-pyrazole-3-carboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A631494-1G A631494-250MG A631494-5G
77292-954EA 167 USD InStock
77292-954 77292-956 77292-958
4-Chloro-1H-pyrazole-3-carboxaldehyde 98%
4-Chloro-1H-pyrazole-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₄H₃ClN₂O
MW: 130.53 g/mol
MDL Number: MFCD00110641
CAS Number: 623570-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty