Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Batimastat 98%

Supplier: AMBEED, INC
Warning

A633905-10MG A633905-1MG A633905-5MG A633905-50MG
77242-904EA 352 USD InStock
77242-904 77242-906 77242-908 77747-306
Batimastat 98%
Batimastat
Precaución: for Research Use Only


Formula: C₂₃H₃₁N₃O₄S₂
MW: 477.64 g/mol
MDL Number: MFCD00866533
CAS Number: 130370-60-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty