Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-(trifluoromethyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A635193-1G A635193-250MG A635193-5G
77266-992EA 27 USD InStock
77266-992 77266-994 77266-996
4-Methoxy-2-(trifluoromethyl)benzoic acid 98%
4-Methoxy-2-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₇F₃O₃
MW: 220.15
MDL Number: MFCD01091013
CAS Number: 127817-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty