Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-amino-5-fluoro-2-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A637545-100MG A637545-1G A637545-250MG A637545-5G
77267-020EA 6 USD InStock
77267-020 77267-022 77267-024 77267-026
Methyl 3-amino-5-fluoro-2-methylbenzoate 97%
Methyl 3-amino-5-fluoro-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₂
MDL Number: MFCD16036853
CAS Number: 697739-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty