Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium(II) trimethylacetate Dimer 98%

Supplier: AMBEED, INC
Warning

A638316-100MG A638316-250MG A638316-50MG
77254-582EA 229 USD InStock
77254-582 77254-584 77254-586
Rhodium(II) trimethylacetate Dimer 98%
Rhodium(II) trimethylacetate Dimer
Precaución: for Research Use Only


Formula: C₂₀H₃₆O₈Rh₂
MW: 610.32 g/mol
MDL Number: MFCD02094719
CAS Number: 62728-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty