Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methylbenzo[d]isoxazole 96%

Supplier: AMBEED, INC
Warning

A640465-1G A640465-250MG
77267-050EA 445.01 USD InStock
77267-050 77267-052
4-Bromo-3-methylbenzo[d]isoxazole 96%
4-Bromo-3-methylbenzo[d]isoxazole
Precaución: for Research Use Only


Formula: C₈H₆BrNO
MDL Number: MFCD22038944
CAS Number: 1367947-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty