Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CC-115 98+%

Supplier: AMBEED, INC
Warning

A642364-10MG A642364-25MG A642364-50MG A642364-5MG
77242-980EA 56 USD InStock
77242-980 77242-982 77242-984 77242-986 77242-988
CC-115 98+%
CC-115
Precaución: for Research Use Only


Formula: C₁₆H₁₆N₈O
MW: 336.35 g/mol
MDL Number: MFCD29036965
CAS Number: 1228013-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty