Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethyl acetate 60%

Supplier: AMBEED, INC
Danger

A644657-1G A644657-25G A644657-5G
77267-088EA 11 USD InStock
77267-088 77267-090 77267-092
2-Hydroxyethyl acetate 60%
2-Hydroxyethyl acetate
Precaución: for Research Use Only


Formula: C₄H₈O₃
MW: 104.1045 g/mol
MDL Number: MFCD00002864
CAS Number: 542-59-6
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty