Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

WDR5 0103 98%

Supplier: AMBEED, INC
Danger

A651436-100MG A651436-10MG A651436-1MG A651436-25MG A651436-50MG A651436-5MG
77243-026EA 138 USD InStock
77243-026 77243-028 77243-030 77243-032 77243-034 77243-036
WDR5 0103 98%
WDR5 0103
Precaución: for Research Use Only


Formula: C₂₁H₂₅N₃O₄
MDL Number: MFCD07086826
CAS Number: 890190-22-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty