Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',4'-Difluoro-4-hydroxybiphenyl 98%

Supplier: AMBEED, INC
Warning

A656863-1G A656863-250MG A656863-5G
77254-754EA 97 USD InStock
77254-754 77254-756 77254-758
2',4'-Difluoro-4-hydroxybiphenyl 98%
2',4'-Difluoro-4-hydroxybiphenyl
Precaución: for Research Use Only


Formula: C₁₂H₈F₂O
MDL Number: MFCD03414359
CAS Number: 59089-68-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty