Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,5R)-1-(2-(1,3-dioxolan-2-yl)phenyl)-2,5-diethylphospholane 98%

Supplier: AMBEED, INC
Warning

A659401-100MG A659401-1G A659401-250MG A659401-5G
77254-772EA 61 USD InStock
77254-772 77254-774 77254-776 77254-778
(2R,5R)-1-(2-(1,3-dioxolan-2-yl)phenyl)-2,5-diethylphospholane 98%
(2R,5R)-1-(2-(1,3-dioxolan-2-yl)phenyl)-2,5-diethylphospholane
Precaución: for Research Use Only


Formula: C₁₇H₂₅O₂P
MDL Number: MFCD09265098
CAS Number: 1710765-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty