Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Ethynylacetophenone 95%

Supplier: AMBEED, INC
Warning

A660492-100MG A660492-250MG A660492-1G A660492-5G
77293-098EA 10 USD InStock
77293-098 77293-100 77552-016 77749-362
4'-Ethynylacetophenone 95%
4'-Ethynylacetophenone
Precaución: for Research Use Only


Formula: C₁₀H₈O
MDL Number: MFCD04974025
CAS Number: 42472-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty