Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Hydroxy-5'-methylacetanilide 97%

Supplier: AMBEED, INC
Warning

A660742-100G A660742-25G
77254-796EA 31 USD InStock
77254-796 77254-798
2'-Hydroxy-5'-methylacetanilide 97%
2'-Hydroxy-5'-methylacetanilide
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00020183
CAS Number: 6375-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty