Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-4-Bromomandelic acid 95%

Supplier: AMBEED, INC
Warning

A663064-100G A663064-1G A663064-250MG A663064-5G
77243-114EA 102 USD InStock
77243-114 77243-116 77243-118 77243-120
(±)-4-Bromomandelic acid 95%
(±)-4-Bromomandelic acid
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MW: 231.05
Boiling Pt: 344
MDL Number: MFCD00004232
CAS Number: 6940-50-7
Merck Index: 13,01409
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty