Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,6-diethylaniline 97%

Supplier: AMBEED, INC
Warning

A663325-100G A663325-10G A663325-25G
77280-596EA 143 USD InStock
77280-596 77280-598 77280-600 77280-602
3-Chloro-2,6-diethylaniline 97%
3-Chloro-2,6-diethylaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClN
MDL Number: MFCD00074880
CAS Number: 67330-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty