Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Syringic acid 96%

Supplier: AMBEED, INC
Warning

A663784-100G A663784-1G A663784-5G A663784-25G
77254-816EA 85 USD InStock
77254-816 77254-818 77254-820 77747-352
Syringic acid 96%
Syringic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 351
MDL Number: MFCD00002552
CAS Number: 530-57-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty