Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Aminopyridin-4-yl)acetonitrile 97%

Supplier: AMBEED, INC
Danger

A667065-100MG A667065-1G A667065-250MG
77243-172EA 81 USD InStock
77243-172 77243-174 77243-176
2-(3-Aminopyridin-4-yl)acetonitrile 97%
2-(3-Aminopyridin-4-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₇H₇N₃
MDL Number: MFCD16608700
CAS Number: 1227512-30-2
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty