Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Benzo[d]isoxazol-3-yl)-4-methylbenzamide 98+%

Supplier: AMBEED, INC
Warning

A667119-10MG A667119-1MG A667119-5MG
77243-182EA 25 USD InStock
77243-182 77243-184 77243-186
N-(Benzo[d]isoxazol-3-yl)-4-methylbenzamide 98+%
N-(Benzo[d]isoxazol-3-yl)-4-methylbenzamide
Precaución: for Research Use Only


Formula: C₁₅H₁₂N₂O₂
MDL Number: MFCD08362411
CAS Number: 891025-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty