Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(2-Bromoethoxy)ethoxy)ethanol 98%

Supplier: AMBEED, INC
Danger

A668037-100MG A668037-1G A668037-250MG A668037-5G A668037-25G
77267-260EA 13 USD InStock
77267-260 77267-262 77267-264 77267-266 77749-374
2-(2-(2-Bromoethoxy)ethoxy)ethanol 98%
2-(2-(2-Bromoethoxy)ethoxy)ethanol
Precaución: for Research Use Only


Formula: C₆H₁₃BrO₃
MDL Number: MFCD18205854
CAS Number: 57641-67-5
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty