Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dimethoxybiphenyl 97%

Supplier: AMBEED, INC
Warning

A668955-100G A668955-25G A668955-5G
77254-866EA 162 USD InStock
77254-866 77254-868 77254-870
2,2'-Dimethoxybiphenyl 97%
2,2'-Dimethoxybiphenyl
Precaución: for Research Use Only


Formula: C₁₄H₁₄O₂
MDL Number: MFCD00067073
CAS Number: 4877-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty