Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-5-Methyl-1,2,3,4-tetrahydronaphthalen-1-amine 95%

Supplier: AMBEED, INC
Warning

A673859-100MG A673859-250MG
77254-918EA 96 USD InStock
77254-918 77254-920
(R)-5-Methyl-1,2,3,4-tetrahydronaphthalen-1-amine 95%
(R)-5-Methyl-1,2,3,4-tetrahydronaphthalen-1-amine
Precaución: for Research Use Only


Formula: C₁₁H₁₅N
MDL Number: MFCD07373970
CAS Number: 1055321-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty