Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1-(2-(4-Methoxystyryl)quinazolin-4-yl)-N3,N3-dimethylpropane-1,3-diamine dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A675515-100MG A675515-25MG A675515-50MG
77280-652EA 188 USD InStock
77280-652 77280-654 77280-656
N1-(2-(4-Methoxystyryl)quinazolin-4-yl)-N3,N3-dimethylpropane-1,3-diamine dihydrochloride 98%
N1-(2-(4-Methoxystyryl)quinazolin-4-yl)-N3,N3-dimethylpropane-1,3-diamine dihydrochloride
Precaución: for Research Use Only


Formula: C₂₂H₂₈Cl₂N₄O
MDL Number: MFCD11519956
CAS Number: 1217195-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty