Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,4-dihydroxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A675622-1G A675622-250MG A675622-5G
77267-304EA 44 USD InStock
77267-304 77267-306 77267-308
5-Bromo-2,4-dihydroxybenzaldehyde 97%
5-Bromo-2,4-dihydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅BrO₃
MDL Number: MFCD20133738
CAS Number: 116096-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty