Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Phenyl-1-propanol 98%

Supplier: AMBEED, INC
Warning

A678978-100G A678978-25G A678978-500G A678978-5G
77280-684EA 86 USD InStock
77280-684 77280-686 77280-688 77280-690
(±)-2-Phenyl-1-propanol 98%
(±)-2-Phenyl-1-propanol
Precaución: for Research Use Only


Formula: C₆H₅CH(CH₃)CH₂OH
MDL Number: MFCD00004736
CAS Number: 1123-85-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty