Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-ethoxypyridine 97%

Supplier: AMBEED, INC
Warning

A680038-10G A680038-25G A680038-5G
77267-358EA 52 USD InStock
77267-358 77267-360 77749-398
2-Chloro-6-ethoxypyridine 97%
2-Chloro-6-ethoxypyridine
Precaución: for Research Use Only


Formula: C₇H₈ClNO
MDL Number: MFCD00023465
CAS Number: 42144-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty