Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Dichlorophenyl)acetone 98%

Supplier: AMBEED, INC
Warning

A680141-1G A680141-250MG A680141-5G
77280-704EA 94 USD InStock
77280-704 77280-706 77749-400
(3,4-Dichlorophenyl)acetone 98%
(3,4-Dichlorophenyl)acetone
Precaución: for Research Use Only


Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
MDL Number: MFCD00210406
CAS Number: 6097-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty