Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Bromo-3'-fluoroacetanilide 95%

Supplier: AMBEED, INC
Warning

A681223-1G A681223-25G A681223-5G
77293-140EA 13 USD InStock
77293-140 77293-142 77293-144
4'-Bromo-3'-fluoroacetanilide 95%
4'-Bromo-3'-fluoroacetanilide
Precaución: for Research Use Only


Formula: H₃CC(O)NH(C₆H₃BrF)
MDL Number: MFCD00672937
CAS Number: 351-30-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty