Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-N-methoxy-N-methylbenzenecarboxamide 95%

Supplier: AMBEED, INC
Warning

A682205-100MG A682205-250MG A682205-5G
77293-146EA 10 USD InStock
77293-146 77293-148 77293-150
4-Cyano-N-methoxy-N-methylbenzenecarboxamide 95%
4-Cyano-N-methoxy-N-methylbenzenecarboxamide
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD07367975
CAS Number: 116332-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty