Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2-methoxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A682808-100MG A682808-1G A682808-250MG A682808-5G
77280-732EA 10 USD InStock
77280-732 77280-734 77280-736 77280-738
3-Hydroxy-2-methoxybenzaldehyde 98%
3-Hydroxy-2-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₃
MDL Number: MFCD06656119
CAS Number: 66495-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty