Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,3,4-Trihydroxybenzimidamide 97%

Supplier: AMBEED, INC
Danger

A683669-1G A683669-5G
77243-328EA 37 USD InStock
77243-328 77243-330
N,3,4-Trihydroxybenzimidamide 97%
3,4-Dihydroxybenzamidoxime
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MDL Number: MFCD22413779
CAS Number: 95933-72-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty