Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dipyridyl sulfide 98%

Supplier: AMBEED, INC
Warning

A683890-1G A683890-250MG A683890-5G
77293-170EA 85 USD InStock
77293-170 77293-172 77293-174
4,4'-Dipyridyl sulfide 98%
4,4'-Dipyridyl sulfide
Precaución: for Research Use Only


Formula: C₁₀H₈N₂S
MW: 188.25 g/mol
MDL Number: MFCD00059784
CAS Number: 37968-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty